Product Details
- SNP ID
-
hCV88472417
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241325997 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTTTTTTAATCTTATTTAGCAAC[A/G]GCCAACTCAGTTTATAAATCCAGAA
- Phenotype
-
MIM: 601506
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SEPT2
PubMed Links
Gene Details
- Gene
- SEPT2
- Gene Name
- septin 2
There are no transcripts associated with this gene.
View Full Product Details