Product Details
- SNP ID
-
rs62270307
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:125232388 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGCATCAGGTGAAGATGTCATCC[C/T]AGCTCTATTATCATTTACATTCACC
- Phenotype
-
MIM: 601897
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF148
PubMed Links
Gene Details
- Gene
- ZNF148
- Gene Name
- zinc finger protein 148
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021964.2 |
2824 |
Missense Mutation |
AGG,GGG |
R780G |
NP_068799.2 |
View Full Product Details