Product Details

SNP ID
rs56389920
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:160500221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCCCAAGACAGCCTAGCCTGCA[C/T]TCTACTTGGATAAATTTTACAAGCT
Phenotype
MIM: 602970
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KPNA4 PubMed Links

Gene Details

Gene
KPNA4
Gene Name
karyopherin subunit alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002268.4 3755 UTR 3 NP_002259.1

View Full Product Details