Product Details

SNP ID
rs62346526
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:220499 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACCTTTGCTTTGCTTCTTGGGA[C/T]AATTGTAACAACAATGGCATTCCAT
Phenotype
MIM: 600857
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CCDC127 PubMed Links

Gene Details

Gene
CCDC127
Gene Name
coiled-coil domain containing 127
There are no transcripts associated with this gene.

Gene
SDHA
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001294332.1 Intron NP_001281261.1
NM_004168.3 Intron NP_004159.2
XM_005248331.3 Intron XP_005248388.1
XM_011514072.1 Intron XP_011512374.1
XM_011514073.1 Intron XP_011512375.1
XM_017009685.1 Intron XP_016865174.1

View Full Product Details