Product Details
- SNP ID
-
rs55794957
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:83211305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGTAATAAGAGCTTTACTCTAT[A/G]AGATGTAACAATGACAAAAGAATAT
- Phenotype
-
MIM: 154250
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ME1
PubMed Links
Gene Details
- Gene
- ME1
- Gene Name
- malic enzyme 1
View Full Product Details