Product Details
- SNP ID
-
hCV88688241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:166324535 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTTCACTAGGGTAAGGAAAGACTT[A/G]CCAGGTCATTGACAAGAACTGCAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFT2D1
PubMed Links
Gene Details
- Gene
- SFT2D1
- Gene Name
- SFT2 domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145169.2 |
|
Intron |
|
|
NP_660152.1 |
View Full Product Details