Product Details

SNP ID
rs61759946
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31269382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATCAGAGCCCTGGGCACTGTTG[G/C]TGCCTGGGGTAGAACAAAAAAAAAG
Phenotype
MIM: 142840
Polymorphism
G/C, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-C PubMed Links

Gene Details

Gene
HLA-C
Gene Name
major histocompatibility complex, class I, C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243042.1 1117 Missense Mutation ACC,AGC T351S NP_001229971.1
NM_002117.5 1117 Missense Mutation ACC,AGC T351S NP_002108.4

View Full Product Details