Product Details

SNP ID
rs41551113
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.6:31355319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCCCTCATCCCCCTCCTTACCC[T/C]ATCTCAGGGTGAGGGGCTTCGGCAG
Phenotype
MIM: 142830
Polymorphism
T/C, Transition Substitution
Allele Nomenclature
Literature Links
HLA-B PubMed Links

Gene Details

Gene
HLA-B
Gene Name
major histocompatibility complex, class I, B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005514.7 903 Nonsense Mutation TAG,TGG *298W NP_005505.2
XM_011514557.1 903 Nonsense Mutation TAG,TGG *298W XP_011512859.1
Gene
MIR6891
Gene Name
microRNA 6891
There are no transcripts associated with this gene.

View Full Product Details