Product Details

SNP ID
hCV88759643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35500107 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTGTTGTGCAGTTCTATGAGGC[C/T]CTCCAGCGTCTTGTTCTGCCAGCGC
Phenotype
MIM: 603170 MIM: 602280
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEAD3 PubMed Links

Gene Details

Gene
TEAD3
Gene Name
TEA domain transcription factor 3
There are no transcripts associated with this gene.

Gene
TULP1
Gene Name
tubby like protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289395.1 1254 Missense Mutation AGC,GGC S404G NP_001276324.1

View Full Product Details