Product Details

SNP ID
rs61002683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:45899040 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATTATGCACACTTTGGATCATAC[A/G]TAATTATGAATCGAAAGTATCCTGA
Phenotype
MIM: 607293
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIC5 PubMed Links

Gene Details

Gene
CLIC5
Gene Name
chloride intracellular channel 5
There are no transcripts associated with this gene.

View Full Product Details