Product Details

SNP ID
rs62521906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:97869688 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCCACGGACAGCCGATGGCTCCC[C/T]CTTCCCTCTTCGGAACCTGGTATTT
Phenotype
MIM: 602108
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MATN2 PubMed Links

Gene Details

Gene
MATN2
Gene Name
matrilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317748.1 Intron NP_001304677.1
NM_002380.4 Intron NP_002371.3
NM_030583.3 Intron NP_085072.2
XM_005250920.1 Intron XP_005250977.1
XM_017013417.1 Intron XP_016868906.1
XM_017013418.1 Intron XP_016868907.1

View Full Product Details