Product Details

SNP ID
hCV89011901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104695006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCCTATTGCAAACAGTTCTGAC[A/C]ATGATGTTGCCTTTCTGTGGGAATA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 585 Silent Mutation ACA,ACC T195T NP_001004484.1

View Full Product Details