Product Details

SNP ID
rs56987928
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129831901 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACAGACTGACACTTACATGTC[C/T]GCATCCTCATCCCTTCGGTTGGTTT
Phenotype
MIM: 605204 MIM: 615143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf78 PubMed Links

Gene Details

Gene
C9orf78
Gene Name
chromosome 9 open reading frame 78
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016520.2 411 Silent Mutation GCA,GCG A113A NP_057604.1
XM_011518780.2 411 Intron XP_011517082.1
Gene
TOR1A
Gene Name
torsin family 1 member A
There are no transcripts associated with this gene.

Gene
USP20
Gene Name
ubiquitin specific peptidase 20
There are no transcripts associated with this gene.

View Full Product Details