Product Details

SNP ID
rs61916419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:2970940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGCTGCTTATGCAAGAGAAGAA[A/G]TAAGCTGTACCAGTGCCTCCCATAC
Phenotype
MIM: 601714
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEAD4 PubMed Links

Gene Details

Gene
TEAD4
Gene Name
TEA domain transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003213.3 Intron NP_003204.2
NM_201441.2 Intron NP_958849.1
NM_201443.2 Intron NP_958851.1

View Full Product Details