Product Details

SNP ID
rs59887243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55319133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGGGTGGGAGCTACGAAGTTGCC[A/C]GTTCCCGGTTACTTCCGGGCTTGCG
Phenotype
MIM: 604122
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PIGB PubMed Links

Gene Details

Gene
PIGB
Gene Name
phosphatidylinositol glycan anchor biosynthesis class B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004855.4 62 UTR 5 NP_004846.4
XM_005254795.4 62 Intron XP_005254852.1
XM_011522235.2 62 UTR 5 XP_011520537.1
XM_011522236.2 62 UTR 5 XP_011520538.1
XM_011522237.2 62 Intron XP_011520539.1
XM_017022730.1 62 Intron XP_016878219.1
XM_017022731.1 62 Intron XP_016878220.1
XM_017022732.1 62 Intron XP_016878221.1
Gene
PIGBOS1
Gene Name
PIGB opposite strand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308421.1 62 UTR 5 NP_001295350.1
NM_001308422.1 62 UTR 5 NP_001295351.1
NM_001308423.1 62 UTR 5 NP_001295352.1

View Full Product Details