Product Details

SNP ID
hCV89333463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18093960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCGTGTGGCGCTGATTGGATTTC[A/C]CTCTGTGGGTAAGGTCAGTGATGGT
Phenotype
MIM: 602986
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DRG2 PubMed Links

Gene Details

Gene
DRG2
Gene Name
developmentally regulated GTP binding protein 2
There are no transcripts associated with this gene.

View Full Product Details