Product Details
- SNP ID
-
rs55722259
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:26017021 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCTTTGGGCTAGATGTTTTCAGAT[C/T]ATGCTTAAATAACCGTGGTTAGAAA
- Phenotype
-
MIM: 600192
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SS18
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80099396] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SS18
- Gene Name
- SS18, nBAF chromatin remodeling complex subunit
View Full Product Details