Product Details

SNP ID
rs62191391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:298508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGATCGAGCTGCGCCAGGGGGAG[A/G]GCGGGGTCAGGCACTTGCCAGGGGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C20orf96 PubMed Links

Gene Details

Gene
C20orf96
Gene Name
chromosome 20 open reading frame 96
There are no transcripts associated with this gene.

Gene
ZCCHC3
Gene Name
zinc finger CCHC-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033089.6 946 Missense Mutation AGC,GGC S308G NP_149080.2

View Full Product Details