Product Details

SNP ID
rs59539985
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21712496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTATTGAGAGTTTGTTATATTGG[A/C]TATCAACTCAGAGTTAGGGGCTGGC
Phenotype
MIM: 300226 MIM: 300066
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMPX PubMed Links

Gene Details

Gene
SMPX
Gene Name
small muscle protein, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014332.2 Intron NP_055147.1

View Full Product Details