Product Details

SNP ID
hCV89656835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:19353140 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTTGCGCCACCCGCAGTGCCGCA[C/T]CTGTGGTCTCGCAGGTGGTCTTGCC
Phenotype
MIM: 608891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4757 PubMed Links

Gene Details

Gene
MIR4757
Gene Name
microRNA 4757
There are no transcripts associated with this gene.

Gene
OSR1
Gene Name
odd-skipped related transciption factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145260.2 Intron NP_660303.1

View Full Product Details