Product Details

SNP ID
rs57350646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:98455401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCCTCTGGCAGAATCTGGGCTG[C/T]CCCCTTGTAAGACTTGAAAGCTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A7P1 PubMed Links

Gene Details

Gene
SLC9A7P1
Gene Name
solute carrier family 9 member 7 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details