Product Details
- SNP ID
-
rs55714835
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
17
- Location
-
Chr.1:9148599 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTAACTCCACTCGCTCCAAACTCA[A/G]GTCCAACCATGAAATTATTGATCCC
- Phenotype
-
MIM: 611172
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR34A
PubMed Links
Gene Details
- Gene
- MIR34A
- Gene Name
- microRNA 34a
There are no transcripts associated with this gene.
- Gene
- MIR34AHG
- Gene Name
- MIR34A host gene
There are no transcripts associated with this gene.
View Full Product Details