Product Details
- SNP ID
-
rs59780529
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:127638623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGGTCGGGGGGAGAGGGGCGGGG[C/T]GGAACCGAGGGCGGAGGCCAAGGTG
- Phenotype
-
MIM: 603071
MIM: 607908
MIM: 606541
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR17
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76475129] are located under a probe and SNP(s) [rs78022502] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR17
- Gene Name
- G protein-coupled receptor 17
There are no transcripts associated with this gene.
- Gene
- LIMS2
- Gene Name
- LIM zinc finger domain containing 2
- Gene
- MYO7B
- Gene Name
- myosin VIIB
View Full Product Details