Product Details
- SNP ID
-
hCV89925080
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:61872089 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAGATTAAATGATTACCTTTCGG[A/G]CATTAATGCCTGCAGTTTTTTCTCC
- Phenotype
-
MIM: 605142
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCT4
PubMed Links
Gene Details
- Gene
- CCT4
- Gene Name
- chaperonin containing TCP1 subunit 4
There are no transcripts associated with this gene.
View Full Product Details