Product Details

SNP ID
rs58397615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:4035698 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCTACCCTGGTTTGTAAAATCA[A/G]TTCTGCTACTTATTGGGCATAGAAC
Phenotype
MIM: 607939
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUMF1 PubMed Links

Gene Details

Gene
SUMF1
Gene Name
sulfatase modifying factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164674.1 Intron NP_001158146.1
NM_001164675.1 Intron NP_001158147.1
NM_182760.3 Intron NP_877437.2
XM_011533624.2 Intron XP_011531926.1
XM_011533625.2 Intron XP_011531927.1
XM_011533626.2 Intron XP_011531928.1
XM_017006252.1 Intron XP_016861741.1
XM_017006253.1 Intron XP_016861742.1
XM_017006254.1 Intron XP_016861743.1
XM_017006255.1 Intron XP_016861744.1

View Full Product Details