Product Details

SNP ID
rs55896017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:45083339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGGCATGACTTAAATATCCTAT[C/T]CTCTGGAAAGTGTTCAAGAGGTCTT
Phenotype
MIM: 611735
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDCP1 PubMed Links

Gene Details

Gene
CDCP1
Gene Name
CUB domain containing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022842.4 4945 UTR 3 NP_073753.3
NM_178181.2 4945 Intron NP_835488.1
XM_011534024.1 4945 UTR 3 XP_011532326.1
XM_017007070.1 4945 UTR 3 XP_016862559.1

View Full Product Details