Product Details

SNP ID
rs56400532
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48466994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACCTGCTCCTAGCCTTCCTGCG[A/G]CGCCAGCCACAGCCCTGGTGCCTGG
Phenotype
MIM: 606605 MIM: 607290 MIM: 606609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATRIP PubMed Links

Gene Details

Gene
ATRIP
Gene Name
ATR interacting protein
There are no transcripts associated with this gene.

Gene
SHISA5
Gene Name
shisa family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272065.1 1475 Intron NP_001258994.1
NM_001272066.1 1475 Intron NP_001258995.1
NM_001272067.1 1475 Intron NP_001258996.1
NM_001272068.1 1475 Intron NP_001258997.1
NM_001272082.1 1475 Intron NP_001259011.1
NM_001272083.1 1475 Intron NP_001259012.1
NM_016479.4 1475 Intron NP_057563.3
XM_005265205.1 1475 Intron XP_005265262.1
XM_006713188.2 1475 Intron XP_006713251.1
XM_011533796.1 1475 Intron XP_011532098.1
Gene
TREX1
Gene Name
three prime repair exonuclease 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007248.3 1475 Silent Mutation CGA,CGG R103R NP_009179.2
NM_016381.5 1475 Silent Mutation CGA,CGG R168R NP_057465.1
NM_033629.4 1475 Silent Mutation CGA,CGG R113R NP_338599.1

View Full Product Details