Product Details
- SNP ID
-
rs56252856
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:37104598 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCATAGCCCTTAGTCCTTGCCAC[A/G]GCAGGCTGGTTCTATTAAAATACAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CSNK1A1L
PubMed Links
Gene Details
- Gene
- CSNK1A1L
- Gene Name
- casein kinase 1 alpha 1 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145203.5 |
1067 |
Missense Mutation |
CCG,CTG |
P220L |
NP_660204.2 |
View Full Product Details