Product Details

SNP ID
rs60816799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:113936329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTACAGTGAGGAAATGCAGCAAT[A/T]GAGAAGCTTCTAAATGCTGTAATCC
Phenotype
MIM: 605164
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HDAC2 PubMed Links

Gene Details

Gene
HDAC2
Gene Name
histone deacetylase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001527.3 6483 UTR 3 NP_001518.3
XM_011535788.1 6483 Intron XP_011534090.1
XM_017010799.1 6483 Intron XP_016866288.1

View Full Product Details