Product Details

SNP ID
rs57261684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:44924867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATGGATAACAATTTTTCTCTCTG[A/C]AGAGATGTAAACTGGATTTTTATTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KLHL28 PubMed Links

Gene Details

Gene
KLHL28
Gene Name
kelch like family member 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308112.1 6097 UTR 3 NP_001295041.1
NM_017658.4 6097 UTR 3 NP_060128.2
XM_005267770.3 6097 UTR 3 XP_005267827.1
XM_006720174.3 6097 Intron XP_006720237.1
XM_011536847.2 6097 UTR 3 XP_011535149.1
XM_011536849.2 6097 UTR 3 XP_011535151.1
XM_017021378.1 6097 Intron XP_016876867.1

View Full Product Details