Product Details

SNP ID
rs56972888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57564135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGTGCATTCAAAATATGTCCCTCT[A/C]TAAGCCAGTGGTATAGACACTGTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SLC35F4 PubMed Links

Gene Details

Gene
SLC35F4
Gene Name
solute carrier family 35 member F4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021259.1 Intron XP_016876748.1
XM_017021261.1 Intron XP_016876750.1

View Full Product Details