Product Details

SNP ID
rs55933233
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24037907 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGGAAGATGAAGGACTGGGAGC[A/G]GCTGGTGGCCACAGTGAGCTGCAGG
Phenotype
MIM: 615195
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS4L1 PubMed Links

Gene Details

Gene
DHRS4L1
Gene Name
dehydrogenase/reductase 4 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277864.1 618 Missense Mutation CAG,CGG Q16R NP_001264793.1
XM_006720246.3 618 Intron XP_006720309.1

View Full Product Details