Product Details

SNP ID
rs57072789
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96143259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTTTCCAACCCCTGCCCACAGG[A/G]CTAGGAGGAAGCGGGGGAGGGAGTG
Phenotype
MIM: 608860 MIM: 603068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASTL PubMed Links

Gene Details

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002036.3 1595 Intron NP_001002036.3
XM_011511205.2 1595 Intron XP_011509507.1
XM_011511206.2 1595 Intron XP_011509508.1
XM_011511207.2 1595 Intron XP_011509509.1
XM_011511208.2 1595 Intron XP_011509510.1
Gene
DUSP2
Gene Name
dual specificity phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004418.3 1595 UTR 3 NP_004409.1
XM_017003546.1 1595 UTR 3 XP_016859035.1

View Full Product Details