Product Details

SNP ID
rs61754776
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:43338228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATACCAGCTGCTGTATGCCTACC[C/T]GCGGTAGGTGCTGGACTGTGCCCCA
Phenotype
MIM: 159530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPL PubMed Links

Gene Details

Gene
MPL
Gene Name
MPL proto-oncogene, thrombopoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005373.2 254 Missense Mutation CCG,CTG P70L NP_005364.1
XM_017001320.1 254 Missense Mutation CCG,CTG P127L XP_016856809.1

View Full Product Details