Product Details

SNP ID
rs61783140
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46635947 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAGAAGAGCTGAACTTGGTTGGC[A/G]ATGCACTGTGCCTCAGCAACATTCT
Phenotype
MIM: 608917
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATPAF1 PubMed Links

Gene Details

Gene
ATPAF1
Gene Name
ATP synthase mitochondrial F1 complex assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042546.2 873 Silent Mutation ATC,ATT I227I NP_001036011.2
NM_001243728.1 873 Silent Mutation ATC,ATT I184I NP_001230657.1
NM_001256418.1 873 Silent Mutation ATC,ATT I121I NP_001243347.1
NM_022745.4 873 Silent Mutation ATC,ATT I295I NP_073582.3

View Full Product Details