Product Details

SNP ID
rs61781203
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:32221921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTAGCAACTGCTGGGCCCAGGT[A/C]GGCTCATGAACCCGCTGCAGGCCGG
Phenotype
MIM: 603911
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCDC2B PubMed Links

Gene Details

Gene
DCDC2B
Gene Name
doublecortin domain containing 2B
There are no transcripts associated with this gene.

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003757.3 145 UTR 5 NP_003748.1
XM_017002671.1 145 Intron XP_016858160.1
Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

Gene
TMEM234
Gene Name
transmembrane protein 234
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019118.4 145 Silent Mutation NP_061991.3
XM_011541795.2 145 Silent Mutation XP_011540097.1
XM_017001812.1 145 Silent Mutation XP_016857301.1
XM_017001813.1 145 Silent Mutation XP_016857302.1
XM_017001814.1 145 Silent Mutation XP_016857303.1
XM_017001815.1 145 Silent Mutation XP_016857304.1
XM_017001816.1 145 Silent Mutation XP_016857305.1
XM_017001817.1 145 Silent Mutation XP_016857306.1
XM_017001818.1 145 Silent Mutation XP_016857307.1
XM_017001819.1 145 Silent Mutation XP_016857308.1
XM_017001820.1 145 Silent Mutation XP_016857309.1
XM_017001821.1 145 Silent Mutation XP_016857310.1
XM_017001822.1 145 Silent Mutation XP_016857311.1
XM_017001823.1 145 Silent Mutation XP_016857312.1
XM_017001824.1 145 Silent Mutation XP_016857313.1
XM_017001825.1 145 Silent Mutation XP_016857314.1
XM_017001826.1 145 Silent Mutation XP_016857315.1
XM_017001827.1 145 Silent Mutation XP_016857316.1
XM_017001828.1 145 Silent Mutation XP_016857317.1
XM_017001829.1 145 Silent Mutation XP_016857318.1
XM_017001830.1 145 Silent Mutation XP_016857319.1
XM_017001831.1 145 Intron XP_016857320.1

View Full Product Details