Product Details

SNP ID
rs56699112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:110107112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGCACCCTGAAAGGGTAGCAC[A/G]AGGGAGCTCTTTGTGGTGAGTGGCA
Phenotype
MIM: 602022
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MALL PubMed Links

Gene Details

Gene
MALL
Gene Name
mal, T-cell differentiation protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005434.4 Intron NP_005425.1
XM_011511809.1 Intron XP_011510111.1

View Full Product Details