Product Details

SNP ID
hCV90567424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:7881667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTGCTTCTCTAGGAGGTTGGG[G/T]TTTTTTAAAAAAAGAATTATCTGTG
Phenotype
MIM: 112266 MIM: 616412
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BLOC1S5-TXNDC5 PubMed Links

Gene Details

Gene
BLOC1S5-TXNDC5
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
BMP6
Gene Name
bone morphogenetic protein 6
There are no transcripts associated with this gene.

Gene
TXNDC5
Gene Name
thioredoxin domain containing 5
There are no transcripts associated with this gene.

View Full Product Details