Product Details

SNP ID
rs61845578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14961360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGTTGAACCATTCCACGGGACC[A/G]TCTGTATGCATATAAACCCTAAGTA
Phenotype
MIM: 605988 MIM: 614174
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCLRE1C PubMed Links

Gene Details

Gene
DCLRE1C
Gene Name
DNA cross-link repair 1C
There are no transcripts associated with this gene.

Gene
MEIG1
Gene Name
meiosis/spermiogenesis associated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080836.2 Intron NP_001074305.1

View Full Product Details