Product Details

SNP ID
rs73039834
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:178850282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTGTAGTAGTAAGATCAGTAAAT[G/T]AGAAAAAAAGTAATAATAAATTAGA
Phenotype
MIM: 603874
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ANGPTL1 PubMed Links

Gene Details

Gene
ANGPTL1
Gene Name
angiopoietin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004673.3 2799 UTR 3 NP_004664.1
XM_005245577.2 2799 UTR 3 XP_005245634.1
Gene
RALGPS2
Gene Name
Ral GEF with PH domain and SH3 binding motif 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286247.1 2799 Intron NP_001273176.1
NM_152663.4 2799 Intron NP_689876.2
XM_006711410.3 2799 Intron XP_006711473.1
XM_006711411.3 2799 Intron XP_006711474.1
XM_011509688.1 2799 Intron XP_011507990.1
XM_017001591.1 2799 Intron XP_016857080.1

View Full Product Details