Product Details

SNP ID
rs72885072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165877928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACATTCAAAAAGTAAAGCAATT[C/T]TTAAGATGTCATGCCAAACATACTT
Phenotype
MIM: 612014
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC100506124 PubMed Links

Gene Details

Gene
LOC100506124
Gene Name
uncharacterized LOC100506124
There are no transcripts associated with this gene.

Gene
TTC21B
Gene Name
tetratricopeptide repeat domain 21B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024753.4 5921 Intron NP_079029.3
XM_006712761.1 5921 Intron XP_006712824.1
XM_011511871.2 5921 Intron XP_011510173.1
XM_011511872.2 5921 Intron XP_011510174.1
XM_017004967.1 5921 UTR 3 XP_016860456.1
XM_017004968.1 5921 Intron XP_016860457.1
XM_017004969.1 5921 Intron XP_016860458.1

View Full Product Details