Product Details

SNP ID
rs73048348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:188733738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTGTAAACCCTTTAGTATATGT[A/G]CATTAGTCTGTTTTCACACTCTATA
Phenotype
MIM: 606423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DIRC1 PubMed Links

Gene Details

Gene
DIRC1
Gene Name
disrupted in renal carcinoma 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052952.2 1454 UTR 3 NP_443184.1
XM_017003281.1 1454 Intron XP_016858770.1

View Full Product Details