Product Details

SNP ID
rs73983605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207826173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTACAGTGACAGGGAGCTGGGGG[C/T]GGAGGGAGAAGGCACTTTGCCATGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLEKHM3 PubMed Links

Gene Details

Gene
PLEKHM3
Gene Name
pleckstrin homology domain containing M3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080475.2 4922 UTR 3 NP_001073944.1
XM_005246545.3 4922 UTR 3 XP_005246602.1
XM_006712521.3 4922 Intron XP_006712584.1
XM_011511160.2 4922 UTR 3 XP_011509462.1
XM_011511163.2 4922 Intron XP_011509465.1
XM_017004071.1 4922 Intron XP_016859560.1
XM_017004072.1 4922 UTR 3 XP_016859561.1
XM_017004073.1 4922 UTR 3 XP_016859562.1

View Full Product Details