Product Details

SNP ID
rs73133803
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:83639489 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAATAGAAATTTGTTCCCACTTG[G/T]ATTTTCAACAAAATTATCGGAACAT
Phenotype
MIM: 115435
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HAPLN1 PubMed Links

Gene Details

Gene
HAPLN1
Gene Name
hyaluronan and proteoglycan link protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001884.3 3322 UTR 3 NP_001875.1
XM_011543168.2 3322 UTR 3 XP_011541470.1
XM_017009051.1 3322 UTR 3 XP_016864540.1
XM_017009052.1 3322 UTR 3 XP_016864541.1
XM_017009053.1 3322 UTR 3 XP_016864542.1
XM_017009054.1 3322 UTR 3 XP_016864543.1

View Full Product Details