Product Details

SNP ID
rs73655646
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110874503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAGTAATCATTTTTGCTTTTTTA[C/T]ACATTTCTGCATTTTTCAAGTTTTC
Phenotype
MIM: 602282
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LPAR1 PubMed Links

Gene Details

Gene
LPAR1
Gene Name
lysophosphatidic acid receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001401.3 2397 UTR 3 NP_001392.2
NM_057159.2 2397 UTR 3 NP_476500.1
XM_005251781.3 2397 UTR 3 XP_005251838.1
XM_005251782.3 2397 UTR 3 XP_005251839.1
XM_017014383.1 2397 UTR 3 XP_016869872.1
XM_017014384.1 2397 UTR 3 XP_016869873.1
XM_017014385.1 2397 UTR 3 XP_016869874.1
XM_017014386.1 2397 UTR 3 XP_016869875.1
XM_017014387.1 2397 UTR 3 XP_016869876.1
XM_017014388.1 2397 UTR 3 XP_016869877.1
XM_017014389.1 2397 UTR 3 XP_016869878.1
XM_017014390.1 2397 UTR 3 XP_016869879.1
XM_017014391.1 2397 UTR 3 XP_016869880.1
XM_017014392.1 2397 UTR 3 XP_016869881.1
XM_017014393.1 2397 UTR 3 XP_016869882.1
XM_017014394.1 2397 UTR 3 XP_016869883.1
XM_017014395.1 2397 UTR 3 XP_016869884.1
XM_017014396.1 2397 UTR 3 XP_016869885.1
XM_017014397.1 2397 UTR 3 XP_016869886.1
XM_017014398.1 2397 UTR 3 XP_016869887.1
XM_017014399.1 2397 UTR 3 XP_016869888.1
XM_017014400.1 2397 UTR 3 XP_016869889.1
XM_017014401.1 2397 UTR 3 XP_016869890.1
XM_017014402.1 2397 UTR 3 XP_016869891.1
XM_017014403.1 2397 UTR 3 XP_016869892.1
XM_017014404.1 2397 UTR 3 XP_016869893.1
XM_017014405.1 2397 UTR 3 XP_016869894.1
XM_017014406.1 2397 UTR 3 XP_016869895.1
XM_017014407.1 2397 UTR 3 XP_016869896.1

View Full Product Details