Product Details

SNP ID
rs72795645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58202042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTAGCAAAGGCTGGTTCATAG[A/G]CCCTAGCAGAAGTTTATGGAAAGAC
Phenotype
MIM: 609851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IPMK PubMed Links

Gene Details

Gene
IPMK
Gene Name
inositol polyphosphate multikinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152230.4 Intron NP_689416.1
XM_011539565.2 Intron XP_011537867.1
XM_017016013.1 Intron XP_016871502.1

View Full Product Details