Product Details

SNP ID
rs73436419
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:128794000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCTCCTCCCGGAGGGCCCAGC[A/G]TGCCAGGAGACACGCTGCAGTAAGG
Phenotype
MIM: 615806
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC15A4 PubMed Links

Gene Details

Gene
SLC15A4
Gene Name
solute carrier family 15 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145648.3 2103 UTR 3 NP_663623.1
XM_011537895.1 2103 UTR 3 XP_011536197.1
XM_017018791.1 2103 Intron XP_016874280.1
XM_017018792.1 2103 Intron XP_016874281.1
XM_017018793.1 2103 Intron XP_016874282.1

View Full Product Details