Product Details

SNP ID
rs71584818
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:5044245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGCGGCCAGGCCACAGGGGGAG[A/T]GCTCCAGTGTCCCCCGACGGCTGGG
Phenotype
MIM: 176267
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNA5 PubMed Links

Gene Details

Gene
KCNA5
Gene Name
potassium voltage-gated channel subfamily A member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002234.3 327 Missense Mutation GAG,GTG E33V NP_002225.2

View Full Product Details