Product Details

SNP ID
rs72831330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4436259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTGGGCATCGATCTGTCAGTTA[C/G]CAGCTCAGCATGCCAGAGAGTGGCT
Phenotype
MIM: 611701
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SPNS3 PubMed Links

Gene Details

Gene
SPNS3
Gene Name
sphingolipid transporter 3 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320449.1 Intron NP_001307378.1
NM_182538.4 Intron NP_872344.3
XM_011523723.1 Intron XP_011522025.1
XM_011523724.1 Intron XP_011522026.1
XM_011523725.1 Intron XP_011522027.1
XM_011523726.2 Intron XP_011522028.1
XM_011523728.2 Intron XP_011522030.1
XM_017024345.1 Intron XP_016879834.1

View Full Product Details