Product Details
- SNP ID
-
rs73313813
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:56979975 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTTGATCCTCATGGAACTTATTTC[A/G]TTTATAGTGTTGAGAGGACCTAAAT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SCPEP1
PubMed Links
Gene Details
- Gene
- SCPEP1
- Gene Name
- serine carboxypeptidase 1
View Full Product Details